Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BLMH/Bleomycin Hydrolase Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298334PE
Description
BLMH/Bleomycin Hydrolase Polyclonal antibody specifically detects BLMH/Bleomycin Hydrolase in Mouse samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
BLMH/Bleomycin Hydrolase | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
BHBMHBLM hydrolase, bleomycin hydrolase, EC 3.4.22.40 | |
Produced in rabbits immunized with purified, recombinant Mouse BLMH/Bleomycin Hydrolase (Accession#: NP_848760.1; Asn 2-Glu 455) | |
0.1 mL | |
Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
642 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction