Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BLMH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580378
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human BLMH/Bleomycin Hydrolase.
The normal physiological role of BLM hydrolase is unknown, but it catalyzes the inactivation of the antitumor drug BLM (a glycopeptide) by hydrolyzing the carboxamide bond of its B-aminoalaninamide moiety thus protecting normal and malignant cells from BLM toxicity.
Specifications
BLMH | |
Polyclonal | |
Unconjugated | |
BLMH | |
AI035728; BH; Bleomycin hydrolase; BLM hydrolase; BLMH; BMH | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
642 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q13867 | |
BLMH | |
Recombinant Human BLMH/Bleomycin Hydrolase protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction