Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BLMH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583468 Supplier Invitrogen™ Supplier No. PA583468
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKEPEDGRLV QFLLMNPAND GGQWDMLVNI VEKYGVIPKK CFPESYTTEA TRRMNDILNH KMREFCIRLR NLVHSGATKG EISATQDVMM EEIFRVVCIC LGNP Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 93%.

The normal physiological role of BLM hydrolase is unknown, but it catalyzes the inactivation of the antitumor drug BLM (a glycopeptide) by hydrolyzing the carboxamide bond of its B-aminoalaninamide moiety thus protecting normal and malignant cells from BLM toxicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLMH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BLMH
Gene Accession No. P70645, Q13867, Q8R016
Gene Alias AI035728; BH; Bleomycin hydrolase; BLM hydrolase; BLMH; BMH
Gene Symbols BLMH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BLMH. Recombinant protein control fragment (Product #RP-93200).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104184, 287552, 642
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.