Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BLNK Antibody (SR2143), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32254650UL
Description
BLNK Monoclonal antibody specifically detects BLNK in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
BLNK | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR2143 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
AGM4, B cell linker protein, BASH, B-cell adapter containing a SH2 domain protein, B-cell adapter containing a Src homology 2 domain protein, B-cell linker, B-cell linker protein, BLNK-s, Cytoplasmic adapter protein, LY57, MGC111051, SLP-65BLNK-S, SLP65Ly57, Src homology 2 domain-containing leukocyte protein of 65 kDa | |
A synthesized peptide derived from human BLNK (Uniprot #: Q8WV28) | |
50 μg | |
Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
29760 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction