Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BLNK Monoclonal Antibody (5G9)
GREENER_CHOICE

Catalog No. PIMA515868
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515868 Supplier Invitrogen™ Supplier No. MA515868
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15868 targets BLNK in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human and mouse samples. The MA5-15868 immunogen is purified recombinant fragment of human BLNK expressed in E. Coli. . MA5-15868 detects BLNK which has a predicted molecular weight of approximately 68kDa.

B cell linker protein (Blnk) is a protein encoded by the Blnk gene located on chromosome 10 in humans. It is also known as SLP 65 (Src homology 2 domain-containing leukocyte protein of 65 kDa). It derives its name from being an adaptor protein in the B cell receptor signaling cascade with several supporting studies showing improper development of B cells in the absence of Blnk. Blnk undergoes tyrosine phosphorylation by Syk, a B cell receptor activated kinase, which provides a platform for interaction of signaling intermediaries such as the Vav GEF and Grb2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLNK
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5G9
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene BLNK
Gene Accession No. Q8WV28, Q9QUN3
Gene Alias AGM4; B cell adaptor containing SH2 domain; B cell linker; B cell linker protein; BASH; bca; B-cell activation; B-cell adapter containing a SH2 domain protein; B-cell adapter containing a Src homology 2 domain protein; B-cell linker; B-cell linker protein; Blnk; BLNK-S; Cytoplasmic adapter protein; LY57; Ly-57; lymphocyte antigen 57; Lyw-57; MGC111051; SLP65; Slp-65; Src homology [SH2] domain-containing leukocyte protein of 65 kD; Src homology 2 domain-containing leukocyte protein of 65 kDa
Gene Symbols BLNK
Host Species Mouse
Immunogen Purified recombinant fragment of human BLNK expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17060, 29760
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.