Learn More
BLNK Mouse anti-Human, Clone: OTI4C9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808517

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
B cell linker protein (Blnk) is a protein encoded by the Blnk gene located on chromosome 10 in humans. It is also known as SLP 65 (Src homology 2 domain-containing leukocyte protein of 65 kDa). It derives its name from being an adaptor protein in the B cell receptor signaling cascade with several supporting studies showing improper development of B cells in the absence of Blnk. Blnk undergoes tyrosine phosphorylation by Syk, a B cell receptor activated kinase, which provides a platform for interaction of signaling intermediaries such as the Vav GEF and Grb2.Specifications
BLNK | |
Monoclonal | |
Unconjugated | |
BLNK | |
AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca | |
Mouse | |
Affinity Chromatography | |
RUO | |
29760 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4C9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WV28 | |
BLNK | |
Human recombit protein fragment corresponding to amino acids 1-292 of human BLNK produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.