Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BLOC1S2 Antibody (03), Alexa Fluor™ 647 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306213AF647
Description
BLOC1S2 Monoclonal antibody specifically detects BLOC1S2 in Human samples. It is validated for ELISASpecifications
BLOC1S2 | |
Monoclonal | |
Alexa Fluor 647 | |
biogenesis of lysosomal organelles complex-1, subunit 2, BLOC-1 subunit 2, CEAP, centrosomal 10 kDa protein, centrosome protein oncogene, Centrosome-associated protein, subunit 2 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human BLOC1S2 (Q6QNY1-2; Met1-Arg99). | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
282991 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
03 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction