Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BLOC1S2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300218AF405
Description
BLOC1S2 Polyclonal specifically detects BLOC1S2 in Human samples. It is validated for ELISA.Specifications
BLOC1S2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
282991 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 405 | |
biogenesis of lysosomal organelles complex-1, subunit 2, BLOC-1 subunit 2, CEAP, centrosomal 10 kDa protein, centrosome protein oncogene, Centrosome-associated protein, subunit 2 | |
Produced in rabbits immunized with purified, recombinant BLOC1S2 (Q6QNY1-2; Met1-Arg99). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction