Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BLOC1S2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561647 Supplier Invitrogen™ Supplier No. PA561647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAEGVLATRS DEPARDDAAV ETAEEAKEP Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 86%.

BLOC1S2 is a component of the ubiquitously expressed BLOC1 multisubunit protein complex. BLOC1 is required for normal biogenesis of specialized organelles of the endosomal-lysosomal system, such as melanosomes and platelet dense granules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLOC1S2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BLOC1S2
Gene Accession No. Q6QNY1
Gene Alias 11 kDa centrosome associated protein; 2410089B13Rik; biogenesis of lysosomal organelles complex 1 subunit 2; biogenesis of lysosomal organelles complex-1, subunit 2; biogenesis of lysosome-related organelles complex 1 subunit 2; biogenesis of lysosome-related organelles complex-1, subunit 2; biogenesis of lysosome-related organelles complex-1, subunit 2A; BLOC-1 subunit 2; BLOC1S2; Bloc1s2a; BLOS2; BORCS2; CEAP; CEAP11; centrosomal 10 kDa protein; centrosome protein oncogene; Centrosome-associated protein; RP11-316M21.4
Gene Symbols BLOC1S2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BLOC1S2. Recombinant protein control fragment (Product #RP-100818).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282991
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.