Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BLOC1S2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA525452
Description
This antibody is predicted to react with non-human primate based on sequence homology.
BLOC1S2 is a component of the ubiquitously expressed BLOC1 multisubunit protein complex. BLOC1 is required for normal biogenesis of specialized organelles of the endosomal-lysosomal system, such as melanosomes and platelet dense granules.
Specifications
| BLOC1S2 | |
| Polyclonal | |
| Unconjugated | |
| BLOC1S2 | |
| 11 kDa centrosome associated protein; 2410089B13Rik; biogenesis of lysosomal organelles complex 1 subunit 2; biogenesis of lysosomal organelles complex-1, subunit 2; biogenesis of lysosome-related organelles complex 1 subunit 2; biogenesis of lysosome-related organelles complex-1, subunit 2; biogenesis of lysosome-related organelles complex-1, subunit 2A; BLOC-1 subunit 2; BLOC1S2; Bloc1s2a; BLOS2; BORCS2; CEAP; CEAP11; centrosomal 10 kDa protein; centrosome protein oncogene; Centrosome-associated protein; RP11-316M21.4 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 282991, 73689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6QNY1, Q9CWG9 | |
| BLOC1S2 | |
| KLH conjugated synthetic peptide between 14-40 amino acids from the central region of human BLOC1S2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction