Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BLOC1S3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565597
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565597 Supplier Invitrogen™ Supplier No. PA565597
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPPLVVQRESA EEAWGTEEAP APAPARSLLQ LRLAES Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.

Component of the BLOC-1 complex, a complex that is required for normal biogenesis of lysosome-related organelles (LRO), such as platelet dense granules and melanosomes. In concert with the AP-3 complex, the BLOC-1 complex is required to target membrane protein cargos into vesicles assembled at cell bodies for delivery into neurites and nerve terminals. The BLOC-1 complex, in association with SNARE proteins, is also proposed to be involved in neurite extension. Plays a role in intracellular vesicle trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLOC1S3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BLOC1S3
Gene Accession No. Q6QNY0
Gene Alias biogenesis of lysosomal organelles complex 1 subunit 3; biogenesis of lysosomal organelles complex-1, subunit 3; biogenesis of lysosome-related organelles complex 1 subunit 3; biogenesis of lysosome-related organelles complex-1, subunit 3; BLOC-1 subunit 3; BLOC1S3; BLOS3; HPS8; reduced pigmentation, mouse, homolog of; RP
Gene Symbols BLOC1S3
Host Species Rabbit
Immunogen Recombinant Human BLOC1S3. Recombinant protein control fragment (Product #RP-106259).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388552
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.