Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blonanserin 98.0+%, TCI America™
SDP

Catalog No. B4565100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4565100MG 100 mg
B456525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4565100MG Supplier TCI America Supplier No. B4565100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 132810-10-7
Molecular Formula C23H30FN3
Molecular Weight (g/mol) 367.51
MDL Number MFCD00893838
InChI Key XVGOZDAJGBALKS-UHFFFAOYSA-N
Synonym 2-(4-Ethyl-1-piperazinyl)-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridine
PubChem CID 125564
IUPAC Name 1-ethyl-4-[4-(4-fluorophenyl)-5H,6H,7H,8H,9H,10H-cycloocta[b]pyridin-2-yl]piperazine
SMILES CCN1CCN(CC1)C1=CC(C2=CC=C(F)C=C2)=C2CCCCCCC2=N1

Specifications

Melting Point 124°C
Color White
Quantity 100 mg
Formula Weight 367.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Blonanserin
RTECSNumber GY0175140
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.