Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Blood 3D Model Making Kit
SCI_ED

Catalog No. S436050 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S436050 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S436050 Supplier NewPath Learning™ Supplier No. 247780
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Blood 3D Model Making Kit allows students to make models of the different components of blood to look at the functions of red blood cells, white blood cells, platelets and plasma.

  • Our blood is a tissue that consists of several different cell types working together.
  • It transports nutrients and oxygen and removes waste products such as carbon dioxide and urea from our tissues and organs.
  • Blood also carries hormones from glands to the tissues where they will function.
  • First, your students will make models out of the colorful parts of the different components of blood.
  • Then they will use their 3D models to look at the functions of red blood cells, white blood cells, platelets and plasma.
  • The model also shows how hemoglobin transports oxygen from the lungs to our organs and how scabs form at wounds.
  • The Model Making Kit includes enough materials for students to assemble up to 5 paper models working cooperatively.
  • The comprehensive Teacher Resource Guide provides illustrated readings, activities, key vocabulary and an assessment, along with teaching opportunities for students to expand their understanding of the concepts presented.

Specifications

For Use With (Application) Model Assembly
Grade 6 to 10
Includes 5 Models, Teacher Resource Guide, Vinyl Pouch
Length (English) 12 in.
Material Paper
Weight (English) 2 lb.
Width (English) 9 in.
Product Type 3D Model
Grade Level Intermediate - Middle School, High School
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.