Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Blood Agar (TSA with Sheep Blood) Medium

Catalog No. r01204
Click to view available options
Quantity:
10/Pk.
100/Pk.
15/Pk.
150/Pk.
20/Pk.
Format:
10mm x 100mm Spacesaver Plate
10mm x 85mm Spacesaver plate
12mm x 85mm Monoplate
15mm x 103mm Tube
85mm Monoplate
Tube
Includes:
TSA w/10% Sheep Blood
TSA w/5% Sheep Blood
Manufacturing Origin:
United States
This item is not returnable. View return policy
This item is not returnable. View return policy

Grow a wide variety of microorganisms on this general purpose non-selective medium that includes sheep blood to promote hemolysis.

Grow a wide variety of fastidious organisms on Thermo Scientific™ Blood Agar (TSA with Sheep Blood), a general purpose non-selective medium with the addition of sheep blood in the formulation to promote the presence and visualization of haemolytic reactions so it can be used as a diagnostic tool.

Tryptone Soya Agar (casein soya bean digest agar) with defibrinated sheep blood is suitable for the cultivation of fastidious aerobic and anaerobic bacteria, the latter of which can be grown either in deep cultures or by incubation under anaerobic conditions. The combination of casein digest and papaic digest of soybean meal leads to optimal growth due to synergy between the protein supply of casein and the carbohydrate supply of soybeans. Sterile defibrinated sheep blood used to enrich the medium produces the hemolysis characteristics of different bacteria, such as streptococci, listeria, hemolytic Staphylococcus, Escherichia coli and Pseudomonas.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Format 12mm x 85mm Monoplate
Manufacturing Origin United States
Media Type Blood Agar
Product Type Agar
Includes TSA w/10% Sheep Blood
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.