Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Blood Bank Crossmatch Holders
SCI_ED

Catalog No. S29188 Shop All Fisherbrand Products
Click to view available options
Positions:
24
29
38

For typing, screening, and other blood bank applications.

  • Clear acrylic shelves with white bases and sides
  • Holds donor sample tubes as well as tubes for donor's serum and cell suspension, ABO, Rh, antibody grouping, and screening
  • Available in four sizes

24-tube capacity:
Holds four 16 mm O.D. sample tubes and twenty 10 mm O.D. crossmatch tubes

29-tube capacity:
Holds one 16 mm sample tube, one 13 mm pilot tube serum, and twenty-seven 10 mm phenotype, genotype, and cross-match tubes

38-tube capacity:
Holds thirty-eight tubes for five sets of tubes with printed identification on top panel

  • Set 1: One 16 mm patient, one serum tube, and one cell suspension tube
  • Set 2: Four 10 mm-14 mm forwarding group drives
  • Set 3: Three reverse grouping tubes
  • Set 4: Sixteen major cross match tubes
  • Set 5: Twelve minor cross match tubes

90-tube capacity:
Holds six 16 mm O.D. donor sample tubes, eighty-four 10 to 13 mm tubes for donor's serum and cell suspension, ABO, Rh, antibody grouping, and screening

Specifications

Product Type Tube Rack
Autoclavable Not Autoclavable
Color Clear, Transparent, White
Holds Tubes (10 to 16 mm)
Material Acrylic
Positions 29
For Use With (Equipment) Blood bank crossmatch tubes
Dimensions (L x W x H) 8.75 x 2.5 x 2.5 in. (22.22 x 6.35 x 6.35 cm)
Length (Metric) 22.22 cm
Length (English) 8.75 in.
Width (Metric) 6.35 cm
Width (English) 2.5 in.
Height (Metric) 6.35 cm
Height (English) 2.5 in.
Diameter (Metric) Opening 10 to 16 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.