Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Blood Group A1B Antigen Monoclonal Antibody (HE-24)
GREENER_CHOICE

Catalog No. PIMA544037
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544037 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544037 Supplier Invitrogen™ Supplier No. MA544037
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody The antibody HE-24 distinguishes blood group A1B from A2B. The specifity of the antibody HE-24 was verified on >1000 samples of blood.

Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Blood Group A1B Antigen
Applications Agglutination, Flow Cytometry
Classification Monoclonal
Clone HE-24
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 15mM sodium azide; pH 8
Gene Alias Blood Group A
Host Species Mouse
Immunogen Glycoprotein fraction isolated from saliva of secretors with blood group A.
Purification Method Sequential Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.