Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Blood Group ABH Antigen Monoclonal Antibody (Z5H-2/Z2A)
GREENER_CHOICE

Catalog No. 01670401
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-401 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-401 Supplier Invitrogen™ Supplier No. MA17667
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7667 detects AB human blood protein from human samples. MA1-7667 has been successfully used in agglutination applications. The MA1-7667 immunogen is AB human blood protein.

Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Blood Group ABH Antigen
Applications Agglutination
Classification Monoclonal
Clone Z5H-2/Z2A
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1.25mM EDTA, 0.3M NaCl and 0.1% sodium azide; pH 7.2
Gene Alias ABH; BG-H; BG-H1
Host Species Mouse
Immunogen AB human blood protein.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.