Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blood Group Antigen A Mouse anti-Human, Clone: HE-193, Abnova™

Catalog No. 89933649 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-649 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-649 Supplier Abnova Corporation Supplier No. MAB13294
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native human Blood Group Antigen A.

Specifications

Antigen Blood Group Antigen A
Applications Agglutination, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone HE-193
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human Blood Group Antigen A.
Dilution Agglutination Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS.
Host Species Mouse
Immunogen A mixture of erythrocytes of Blood Group Antigen A and glycoprotein fraction isolated from the saliva of secretors with Blood Group Antigen A.
Purification Method PEG precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This monoclonal antibody recognizes human blood group A (monofucosyl and difucosyl A antigens with chain types 1, 2, 3, 4, 5, 6) and Forssmann antigen.
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.