Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Blood Group Antigen H (O) Type 1 Monoclonal Antibody (17-206), eBioscience™, Invitrogen™

Catalog No. 14981037
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-981-037 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-981-037 Supplier Invitrogen™ Supplier No. 14981037
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MAb 17-206 recognizes the human blood group antigen H (O) type 1, a subtype primarily found in epithelial and secretory cells. The precursor H antigen is modified by addition of saccharides to carbohydrate chains of proteins and lipids on erythrocytes and epithelial cells. The type, attachment, location, and sequence of saccharides determines the blood group antigen subtype and is mediated by multiple glycosyl transferases. Depending on ABO blood type, the H antigen is converted into either the A antigen, B antigen, both the A and B antigen, or left unconverted (O antigen). Evidence that the blood group antigen H is not expressed in tumor cells and that the level correlates with disease progression suggests that the blood group antigen H may be involved in cell proliferation, cell adhesion, and angiogenesis, although its exact function remains to be determined. The 17-206 antibody does not recognize blood group antigen H (O) type 2.

The H antigen is a precursor to the ABO blood groupantigens, present in people of all common blood types. The Bombay phenotype (hh) does not express antigen H on red blood cells, and therefore this type will also lack A or B antigens, similar to the O blood group. However, unlike O group, the H antigen is absent, hence the individuals produce isoantibodies to antigen H as well as to both A and B antigens. If they receive blood from someone with O blood group, the anti-H antibodies will bind to the H antigen on the red blood cells of the donor blood and destroy the RBCs bycomplement-mediated lysis. Therefore, people with Bombay phenotype can receive blood only from otherhhdonors, although they can donate as though they were type O. Some individuals with the blood group A1 may also be able to produce anti-H antibodies due to the complete conversion of all the H antigen to A1 antigen. Production of the H antigen, or its deficiency in the Bombay phenotype, is controlled at the H locus on chromosome 19. The H locus contains threeexonsthat span more than 5kbof genomic DNA, and encodes thefucosyltransferasethat produces the H antigen on RBCs. The H antigen is a carbohydrate sequence withcarbohydrateslinked mainly to protein (with a minor fraction attached toceramidemoiety).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Blood Group Antigen H (O) Type 1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 17-206
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias ABH; BG-H; BG-H1; group h
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.