Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blood Group Antigen H Type 2 Mouse anti-Human, Clone: 19-OLE, Abnova™

Catalog No. 89936702 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-936-702 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-936-702 Supplier Abnova Corporation Supplier No. MAB13291
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native human Blood Group Antigen H Type 2.

Specifications

Antigen Blood Group Antigen H Type 2
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone 19-OLE
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human Blood Group Antigen H Type 2.
Dilution Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS (0.05% BSA, 0.05% sodium azide).
Host Species Mouse
Immunogen Mucinous colonic adenocarcinoma.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.