Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Blood Group H Antigen Monoclonal Antibody (89-8)
GREENER_CHOICE

Catalog No. 01670408
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-408 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-408 Supplier Invitrogen™ Supplier No. MA17674
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7674 detects H1 and H2 human blood from human samples. MA1-7674 has been successfully used in ELISA and agglutination applications. The MA1-7674 immunogen is a H human blood protein. For short term storage, store product at 4ºC. For long-term storage, store at -80ºC.

The H antigen is a precursor to the ABO blood groupantigens, present in people of all common blood types. The Bombay phenotype (hh) does not express antigen H on red blood cells, and therefore this type will also lack A or B antigens, similar to the O blood group. However, unlike O group, the H antigen is absent, hence the individuals produce isoantibodies to antigen H as well as to both A and B antigens. If they receive blood from someone with O blood group, the anti-H antibodies will bind to the H antigen on the red blood cells of the donor blood and destroy the RBCs bycomplement-mediated lysis. Therefore, people with Bombay phenotype can receive blood only from otherhhdonors, although they can donate as though they were type O. Some individuals with the blood group A1 may also be able to produce anti-H antibodies due to the complete conversion of all the H antigen to A1 antigen. Production of the H antigen, or its deficiency in the Bombay phenotype, is controlled at the H locus on chromosome 19. The H locus contains threeexonsthat span more than 5kbof genomic DNA, and encodes thefucosyltransferasethat produces the H antigen on RBCs. The H antigen is a carbohydrate sequence withcarbohydrateslinked mainly to protein (with a minor fraction attached toceramidemoiety).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Blood Group H Antigen
Applications Agglutination, ELISA
Classification Monoclonal
Clone 89-8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 7.2
Gene Alias ABH; BG-H; BG-H1; group h
Host Species Mouse
Immunogen H human blood protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.