Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precipio Bloodhound ABL1 Resistance Comprehensive Validation Set

Catalog No. 22000146
Change view
Click to view available options
Validation Set with Pre-Plated ABL1 Resistance Panel Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-000-146 Validation Set with Pre-Plated ABL1 Resistance Panel Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-000-146 Supplier Precipio Supplier No. BHVALABL1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Validation Set contains 20 blinded control samples (at least 3 mutant, 3 normal alleles per gene), pre-plated ABL1 Resistance panel plates and ABL1 Resistance LoD plate.

  • Results are verified with a key for panel accuracy, sample positive percent agreement (PPA), and negative percent agreement (NPA).

Specifications

Detection Method qPCR
Form Frozen
GC-Rich PCR Performance Medium
PCR Method qPCR
Reaction Speed Standard
Label or Dye Unlabeled
Product Type Comprehensive Validation Set
For Use With (Application) RT-PCR, Hot-start PCR, DNA Quantitation, Oncology, Multiplex PCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.