Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blooms Syndrome Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517222
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517222 Supplier Thermo Scientific Supplier No. PA517222
Add to Cart
Add to Cart

Blooms Syndrome Polyclonal Antibody

Blooms Syndrome Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Blooms Syndrome
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene BLM
Gene Accession No. P54132
Gene Alias Blm; Bloom syndrome homolog; Bloom syndrome protein; Bloom syndrome protein homolog; Bloom syndrome RecQ like helicase; Bloom syndrome, RecQ helicase-like; BS; DNA helicase, RecQ-like type 2; mBLM; MGC126616; MGC131618; MGC131620; RecQ helicase homolog; recQ protein-like 3; RecQ2; RECQL2; RECQL3
Gene Symbols BLM
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the amino terminus of human BLM.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 641
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.