Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Blooms Syndrome Protein Blm Antibody, Alexa Fluor™ 405, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100324AF405
Description
Blooms Syndrome Protein Blm Polyclonal antibody specifically detects Blooms Syndrome Protein Blm in Human samples. It is validated for Western BlotSpecifications
Blooms Syndrome Protein Blm | |
Polyclonal | |
Western Blot | |
Bloom syndrome, Bloom syndrome, RecQ helicase-like, BSBloom syndrome protein, DNA helicase, RecQ-like type 2, EC 3.6.1, EC 3.6.4.12, MGC126616, MGC131618, RecQ protein-like 3, RecQ2, RECQ2MGC131620, RECQL3RECQL2 | |
The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human Bloom Syndrome using the numbering given in entry NP_000048.1 (GeneID 641). | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Genes Sensitive to DNA Damaging Agents, Phospho Specific, Stem Cell Markers, Tumor Suppressors | |
641 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction