Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Blooms Syndrome Protein Blm Antibody (BFL 103), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250122AF350
Description
Blooms Syndrome Protein Blm Monoclonal specifically detects Blooms Syndrome Protein Blm in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Blooms Syndrome Protein Blm | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
BLM | |
Recombinant Full Length Bloom′s Protein | |
0.1 mL | |
641 | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
BFL 103 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Bloom syndrome, Bloom syndrome, RecQ helicase-like, BSBloom syndrome protein, DNA helicase, RecQ-like type 2, EC 3.6.1, EC 3.6.4.12, MGC126616, MGC131618, RecQ protein-like 3, RecQ2, RECQ2MGC131620, RECQL3RECQL2 | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction