Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Blooms Syndrome Protein Blm Antibody (BFL 103) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Blooms Syndrome Protein Blm |
---|---|
Clone | BFL 103 |
Concentration | 0.9 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
Description
Blooms Syndrome Protein Blm Monoclonal antibody specifically detects Blooms Syndrome Protein Blm in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
Blooms Syndrome Protein Blm | |
0.9 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Genes Sensitive to DNA Damaging Agents, Stem Cell Markers, Tumor Suppressors | |
PBS | |
641 | |
IgG1 | |
Protein A purified |
BFL 103 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Bloom syndrome, Bloom syndrome, RecQ helicase-like, BSBloom syndrome protein, DNA helicase, RecQ-like type 2, EC 3.6.1, EC 3.6.4.12, MGC126616, MGC131618, RecQ protein-like 3, RecQ2, RECQ2MGC131620, RECQL3RECQL2 | |
Recombinant Full Length Bloom's Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title