Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Blot Module for miniVE Vertical Electrophoresis System

Catalog No. 45001976 Shop All Cytiva Products
Change view
Click to view available options
Type:
Blot Module
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
45-001-976 Blot Module
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-976 Supplier Cytiva Supplier No. 80641896

missing translation for 'validMainframeMsg'

Includes

3 Dacron sponges 6mm (0.25 in. thick), 25 sheets of blotter paper

Performs 1-D and 2-D electrophoresis and electrotransfer in a single compact unit

  • Can Cast, run, and blot mini-gels in four streamlined components: gel module, blot module, lower buffer chamber, safety lid
  • Can run two mini-gels or four mini-blots at once, using 10 × 10.5cm gel cassettes for increased separation distances
  • Complete semi-wet blotting in 45min with only 300ml buffer
  • Precast gels can be used from a variety of manufacturers
  • Perform two second-dimension separations using 7cm IPG strips in 1.5 hour
  • Power requirement 15w per blot module
  • Working temperature is 5° to 75°C

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
For Use With (Equipment) MiniVE™ Vertical Electrophoresis System
Type Blot Module
Includes 3 Dacron sponges 6mm (0.25 in. thick), 25 sheets of blotter Paper, supplied with blot module
Temperature (English) Operating 41°F to 167°F
Temperature (Metric) Operating 5°C to 75°C
Length (English) Gel 4.1 in.
Length (Metric) Gel 10.5 cm
Width (English) Gel 3.93 in.
Width (Metric) Gel 10 cm
Wattage 15 w
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.