Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IBI Scientific™ Blot Washer

Catalog No. 15453250 Shop All Stovall Life Science Inc. Products
Change view
Click to view available options
Voltage:
115 V
220 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
15-453-250 115 V
15-453-251 220 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-453-250 Supplier IBI Scientific™ Supplier No. BLWAA115S
Add to Cart
Add to Cart

Includes

Pump/control module, 4L wash bottle w/cap, 4L waste bottle with cap, mini blot tray, standard blot tray, vacuum tube with connectors, dispense manifold, aspirate manifold, power cord, and operation manual

For automated dispensing and aspiration of western blot reagents

  • Used with any existing shaking or rocking device
  • Pump/control unit with microprocessor and software for development of 20 stored programs
  • Module contains three solenoid valves, two for delivery and one for aspiration
  • Software allows control of all elements of instruments
  • 20+ liquid delivery and aspiration programs can be programmed
  • Wash time, liquid dispense rate/volume and aspiration rate/volume can be set-up
  • Base Footprint: 9 x 9.75 in. (22.8 x 24.7cm)
  • Overall Height: 5.5 in. (13.9cm)

BLWAA115S

  • Electrical requirement: 115V, 50/60Hz
BLWAA220S
  • Electrical requirement: 220V, 50/60Hz

Pump for liquid delivery pressure is contained in console. Attachment for house vacuum to aspirate liquids in on rear side of console.

Automate the washing process of Western Transfer membranes

Certifications

CE marked (BLWAA220S)

Western Membranes

Specifications

For Use With (Equipment) Shaking or Rocking Device
Type Blot Washer
Includes Pump/control module, 4L wash bottle w/cap, 4L waste bottle with cap, mini blot tray, standard blot tray, vacuum tube with connectors, dispense manifold, aspirate manifold, power cord, and operation manual
Height (English) 5.5 in.
Height (Metric) 13.9 cm
Electrical Requirements 115 V 50/60 Hz
Voltage 115 V
Wattage 10 w
Frequency 50/60 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.