Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLTR-1 (Leukotriene B4 Receptor) Mouse anti-Human, BV750, Clone: 203/14F11 (also known as 14F11), BD Biosciences™

Catalog No. BDB747015 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB747015 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB747015 Supplier BD Biosciences Supplier No. 747015
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BLTR-1 (Leukotriene B4 Receptor) Monoclonal antibody specifically detects BLTR-1 (Leukotriene B4 Receptor) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLTR-1 (Leukotriene B4 Receptor)
Applications Flow Cytometry
Classification Monoclonal
Clone 203/14F11 (also known as 14F11)
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 750
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias hBLTR-1; BLT1; BLTR; P2Y7; GPR16; LTBR1; P2RY7; CMKRL1; LTB4R1
Host Species Mouse
Immunogen Human BLTR-1-transfected cell line
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.