Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLTR-1 Mouse, anti-Human, R718, Clone: 203/14F11, BD OptiBuild™

Catalog No. BDB751954 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751954 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751954 Supplier BD Biosciences Supplier No. 751954
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody 203/14F11 specifically recognizes human leukotriene B4 receptor 1 (BLTR-1). BLTR-1 is a seven-transmembrane, G-protein-coupled receptor. The receptor plays an important role in pro-inflammatory responses. Leukotrienne B4 receptors are distributed in various hematopoietic cells including monocytes, granulocytes, and lymphocytes. Leukotriene B4 (LTB4), a product of arachidonic acid metabolism, is the ligand for BLTR-1. The immunogen used to generate 203/14F11 hybridoma was a human BLTR-1-transfected cell line.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BLTR-1
Applications Flow Cytometry
Classification Monoclonal
Clone 203/14F11
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.