Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Werner Co. Blue Armor Climbing/Positioning Harness with Back, Hip and Front D-Rings Harness

Catalog No. 17103844
Change view
Click to view available options
Material:
Polyester
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
17-103-844 Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-103-844 Supplier Werner Co. Supplier No. H162002
Add to Cart
Add to Cart

Blue armor climbing/positioning harness with back, hip, and front D-rings.

Built in relief handles provide immediate relief after a fall arrest allowing you to reach a gravity override seated position.

  • WebAlert™ inspectable webbing makes wear and damage visible and obvious, and with multiple impact indicators, makes inspection easier.
  • Lightweight, form fitting padding with breathable compression interior is designed to protect user and prevent tangling when donning.
  • Hip D-rings for positioning applications.
  • Front D-ring for climbing applications.
  • Quick connect buckles on chest.

Specifications

Buckle Style (Chest) Quick Connect
Buckle Style (Leg) Tongue Buckle
Material Polyester
Size Medium/Large
Style Blue Armor
Webbing Material Polyester
Color Black, Blue, Yellow
For Use With (Application) Fall Protection
D-Ring Location Back, Chest, Front, Hips, Side
Product Type Crossover-style Positioning/Climbing Harness
Length (Metric) 45.72 cm
Includes Back Padding, Shoulder Padding, Lanyard Keeper
Certifications/Compliance ANSI z359.11-2021,OSHA 29 CFR 1910.140 and 1926.502
Capacity (English) 425 lb.
Weight (English) 6 lb.
Capacity (Metric) 192.78 kg
Length (English) 18 in.
Dimensions (L x W x H) 18 x 14.4 x 2.4 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.