Learn More
R&D Systems™ Blue Counterstain
SDP
Catalog No. 48203013 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mL
Catalog No. Quantity
48-203-013 50 mL
1 options
This item is not returnable. View return policy
Catalog No. 48-203-013 Supplier R&D Systems™ Supplier No. 48203013
Add to Cart
Add to Cart
This item is not returnable. View return policy
R&D Systems™ Blue Counterstain is resuable and comes ready for use.
  • Do not use past expiration date
  • Ready for use
  • May be reused many times
  • Store in a closed container to prevent evaporation
  • If a precipitate forms, filter sample through Whatman 3MM paper
  • Proprietary reagent
Specifications
Quantity 50 mL
Content And Storage Store the unopened product at room temperature
Product Line R&D Systems