Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blue Marker Antibody (6F4-F6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP23337601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23337601 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP23337601 Supplier Novus Biologicals Supplier No. NBP2333760.1ML

Mouse Monoclonal Antibody

Blue Marker Monoclonal specifically detects Blue Marker in Non-species specific samples. It is validated for Western Blot.

Specifications

Antigen Blue Marker
Applications Western Blot
Classification Monoclonal
Clone 6F4-F6
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS with 0.05% Sodium Azide
Gene Alias Blue
Host Species Mouse
Immunogen Blue dye stained proteins
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects BLUE prestained molecular weight markers from different vendors, does not cross-react with whole cell proteins of different species, does not interfere with the detection by other antibodies.
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.