Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nasco Blue Phantom Ultrasound Training Block
SDPSCI_ED

Catalog No. S00098 Shop All Simulaids, Inc. Products
Change view
Click to view available options
For Use With (Application):
Branched 2 Vessel
Branched 4 Vessel
Pediatric 4 Vessel
Regional Anesthesia
Soft Tissue Biopsy
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S00098 Soft Tissue Biopsy
S00200 Pediatric 4 Vessel
S00100 Branched 2 Vessel
S00199 Branched 4 Vessel
S00101 Regional Anesthesia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S00098 Supplier Nasco Supplier No. BPTM130
Add to Cart
Add to Cart

Blue Phantom Ultrasound Training Block provides practice for operating an ultrasound for various applications.

  • Ultrasound training block
  • Blue Phantom Series

Specifications

Product Type Ultrasound Training Block
For Use With (Application) Soft Tissue Biopsy
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.