Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Blue Sepharose™ 6 Fast Flow Chromatography Media

Catalog No. 45002968 Shop All Cytiva Products
Change view
Click to view available options
Particle Size:
45 to 165 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size
45-002-968 45 to 165 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-968 Supplier Cytiva Supplier No. 17094801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blue dye binds to proteins such as albumin and lipoproteins as well as enzymes like kinases and dehydrogenases

This medium is well established as adsorbent for albumin and interferon at laboratory and process scale.

  • Cibacron Blue 3G is covalently coupled to the base matrix allowing for low ligand leakage and high capacity
  • The blue dye binds proteins, such as albumin, interferon, lipoproteins and blood coagulation factors
  • The dye also binds enzymes including kinases, dehydrogenases, and most enzymes requiring adenyl-containing cofactors e.g. NAD+
  • Avg. particle size: 90μm
  • Volume: 50mL

Developed and tested in co-operation with large-scale manufacturers and is used in routine commercial production.

  • For large-scale purification of interferon and albumin
  • For isolating groups of enzymes and removing of albumin
  • For industrial applications and other approved applications

Certifications

Certificate of Analysis

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mL
For Use With (Application) For Albumin and Interferon at Both Laboratory and Process Scale
Stability Stable in commonly used aqueous Buffers: 70% Ethanol, 6 M Guanidine Hydrochloric, 8 M Urea
Storage Requirements 2°C to 8°C
Product Type Blue Sepharose 6 Fast Flow Media
Flow Rate 250 to 400 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Includes Certificate of Analysis
Max. Pressure 100 kPa
Matrix 6% Cross-Linked Agarose
pH Range 4 to 12
Storage Buffer 20% Ethanol, 0.1 M Potassium Phosphate Buffer
Ligand Type Cibacron Blue
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.