Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLVRA Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501567949
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7949 Supplier Bethyl Laboratories, Inc Supplier No. A304527A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Biliverdin reductase (BLVR) is a ubiquitously expressed, small cytoplasmic oxidoreductase that catalyzes the reduction of biliverdin to bilirubin. The first step of this process is catalyzed by heme oxygenase, an enzyme that converts heme to iron, carbon monoxide, and biliverdin. BLVR is unique among all enzymes to date in being dual pH/dual cofactor-dependent, as it requires NADH (pH 6.7) or NADPH (pH 8.7) as an electron donor. There are two isoforms of BLVR, biliverdin-IX alpha-reductase (BLVRA), the major component of human adult liver, and biliverdin-IX beta-reductase (BLVRB), found predomitly in fetal liver. The product of BLVR, bilirubin, is an important biological antioxidant, however it is also a neurotoxin and the cause of kernicterus. In addition to being an antioxidant, it inhibits kinase activity and the activity of enzymes such as protein kinase C and NADPH oxidase.

Specifications

Antigen BLVRA
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene BLVRA
Gene Accession No. P53004
Gene Alias BLVRA, Biliverdin reductase A, EC 1.3.1.24, BVR A
Gene Symbols BLVRA
Host Species Rabbit
Immunogen Between 75 and 125
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.