Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BLVRB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5122197
This item is not returnable.
View return policy
Description
BLVRB Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The final step in heme metabolism in mammals is catalyzed by the cytosolic biliverdin reductase enzymes A and B (EC 1.3.1.24).
Specifications
BLVRB | |
Polyclonal | |
Unconjugated | |
BLVRB | |
Biliverdin reductase B; biliverdin reductase B (flavin reductase (NADPH)); biliverdin-IX beta-reductase; BLVRB; BVRB; BVR-B; epididymis secretory protein Li 10; flavin reductase (NADPH); FLR; FR; GHBP; green heme-binding protein; HEL-S-10; NADPH; NADPH-dependent diaphorase; NADPH-flavin reductase; SDR43U1; short chain dehydrogenase/reductase family 43U, member 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
292737, 645 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P30043 | |
BLVRB | |
Recombinant protein Biliverdin Reductase B. The antigen corresponds to amino acid range 2-206 of the target protein. | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction