Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BLVRB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 177271AP150UL

Description
The final step in heme metabolism in mammals is catalyzed by the cytosolic biliverdin reductase enzymes A and B (EC 1.3.1.24).Specifications
BLVRB | |
Polyclonal | |
Unconjugated | |
BLVRB | |
Biliverdin IX beta reductase, Biliverdin reductase B, BLVRB, BVR B, BVRB, Flavin reductase, FLR, FR, GHBP, Green heme binding protein, NADPH dependent diaphorase, NADPH flavin reductase, SDR43U1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
233016, 292737, 645 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30043, Q923D2 | |
Blvrb | |
BLVRB Fusion Protein Ag12124 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction