Learn More
Invitrogen™ BLZF1 Monoclonal Antibody (2E10)

Description
Peptide Sequence: KLAKAVNSHL LGNVGINNQK KIPSTVEFCS TPAEKMAETV LRILDPVTCK ESSPDNPFFE SSPTTLLATK KNIGRFHPYT RYENITFNCC NHCRGELIAL.
Specifications
Specifications
| Antigen | BLZF1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2E10 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | BLZF1 |
| Gene Accession No. | Q9H2G9 |
| Gene Alias | 1700030G05Rik; Basic leucine zipper nuclear factor 1; basic leucine zipper nuclear factor 1 (JEM-1); Blzf1; blzf1 protein; Blzf1l; cytoplasmic protein; fe06a04; Golgin45; Golgin-45; Golgin-45; golgin-45; JEM1; JEM-1; JEM-1s; JEM-1short protein; LOC504820 protein; MGC22497; p45 basic leucine-zipper nuclear factor; wu:fe06a04; zgc:66280; zgc:85871 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.