Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BLZF1 Monoclonal Antibody (OTI8E8)
GREENER_CHOICE

Catalog No. PIMA527126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527126 Supplier Invitrogen™ Supplier No. MA527126
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BLZF1 Monoclonal Antibody for Western Blot

Required for normal Golgi structure and for protein transport from the endoplasmic reticulum (ER) through the Golgi apparatus to the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLZF1
Applications Western Blot
Classification Monoclonal
Clone OTI8E8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BLZF1
Gene Accession No. Q9H2G9
Gene Alias 1700030G05Rik; Basic leucine zipper nuclear factor 1; basic leucine zipper nuclear factor 1 (JEM-1); Blzf1; blzf1 protein; Blzf1l; cytoplasmic protein; fe06a04; Golgin45; Golgin-45; Golgin-45; golgin-45; JEM1; JEM-1; JEM-1s; JEM-1short protein; LOC504820 protein; MGC22497; p45 basic leucine-zipper nuclear factor; wu:fe06a04; zgc:66280; zgc:85871
Gene Symbols BLZF1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-246 of BLZF1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8548
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.