Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLZF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541466
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541466 Supplier Thermo Scientific Supplier No. PA541466
Add to Cart
Add to Cart

BLZF1 Polyclonal Antibody

BLZF1 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen BLZF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BLZF1
Gene Accession No. Q9H2G9
Gene Alias 1700030G05Rik; Basic leucine zipper nuclear factor 1; basic leucine zipper nuclear factor 1 (JEM-1); Blzf1; blzf1 protein; Blzf1l; cytoplasmic protein; fe06a04; Golgin45; Golgin-45; Golgin-45; golgin-45; JEM1; JEM-1; JEM-1s; JEM-1short protein; LOC504820 protein; MGC22497; p45 basic leucine-zipper nuclear factor; wu:fe06a04; zgc:66280; zgc:85871
Gene Symbols BLZF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human BLZF1 (aa 345-394).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8548
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.