Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMAL1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$223.52 - $601.00

Specifications

Antigen BMAL1
Concentration 1.0 mg/mL
Dilution Western Blot 0.5 ug/mL - 2 ug/mL, Flow Cytometry, Immunohistochemistry 1:250. Use reported in scientific literature (PMID 33510438), Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen reported in scientific literature (PMID 23736292), Flow (Intracellular), Chromatin Immunoprecipitation (ChIP), Knockout Validated
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1002288SS
SDP
View Documents
Novus Biologicals
NB1002288SS
0.025 mL
Each for $223.52
Only null left
Add to Cart
 
NB1002288
SDP
View Documents
Novus Biologicals
NB1002288
0.1 mL
Each for $601.00
Only null left
Add to Cart
 
Description

Description

BMAL1 Polyclonal specifically detects BMAL1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated.
Specifications

Specifications

BMAL1
Western Blot 0.5 ug/mL - 2 ug/mL, Flow Cytometry, Immunohistochemistry 1:250. Use reported in scientific literature (PMID 33510438), Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen reported in scientific literature (PMID 23736292), Flow (Intracellular), Chromatin Immunoprecipitation (ChIP), Knockout Validated
Polyclonal
Rabbit
Cholesterol Metabolism, Chromatin Research, Circadian Rhythm, Hypoxia, Lipid and Metabolism, Neuroscience, Transcription Factors and Regulators, Vision
PBS with 0.02% Sodium Azide
aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, Member of PAS protein 3, member of PAS superfamily 3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3MGC47515
ARNTL
IgG
Affinity Purified
70 kDa
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunofluorescence
Unconjugated
RUO
Human, Mouse, Rat, Primate
O00327
406
Bacterially expressed human BMAL1 (C-terminus). [UniProt# O00327].
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.