Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMAL1 Antibody (OTI1H6), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270208C
Description
BMAL1 Monoclonal specifically detects BMAL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| BMAL1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ARNTL | |
| Full length recombinant protein of human BMAL1 (NP_001169) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1H6 | |
| Western Blot, Immunohistochemistry | |
| aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, Member of PAS protein 3, member of PAS superfamily 3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3MGC47515 | |
| Mouse | |
| Affinity Purified | |
| Cholesterol Metabolism, Chromatin Research, Circadian Rhythm, Hypoxia, Lipid and Metabolism, Neuroscience, Transcription Factors and Regulators, Vision | |
| 406.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction