Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMAL1 Guinea Pig anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE259925UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2599-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE2599-25UL Supplier MilliporeSigma Supplier No. ABE259925UL
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Anti-BMAL1 Antibody is a highly specific guinea pig polyclonal antibody, that targets Aryl hydrocarbon receptor nuclear translocator-like protein 1 and has been tested in Immunoprecipitation and ChIP-seq and Chromatin Immunoprecipitation (ChIP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMAL1
Applications ChIP Assay
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Arnt1,Bmal1
Host Species Guinea Pig
Immunogen KLH-conjugated linear peptide corresponding to 37 amino acids from the N-terminal half of murine Aryl hydrocarbon receptor nuclear translocator-like protein 1 (BMAL1). This sequence is conserved in isoforms 2, 4, and 5.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity This guinea-pig polyclonal antibody detects Aryl hydrocarbon receptor nuclear translocator-like protein 1 (BMAL1). It targets an epitope within 37 amino acids from the N-terminal half.
Target Species Human, Rat, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.