Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMAL1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991592
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1592 25 μL
50-199-1591 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1592 Supplier Rockland Immunochemicals Supplier No. 600101ML8S
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human Bmal1. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to mouse, rat, golden hamster, horse, gallus gallus, Pongo abelii, Tyto alba, and Nannospalax galili.

The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene.

Specifications

Antigen BMAL1
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Arntl
Gene Accession No. O00327, Q9WTL8
Gene Alias Arnt3; Arntl; ARNT-like protein 1, brain and muscle; aryl hydrocarbon receptor nuclear translocator like; aryl hydrocarbon receptor nuclear translocator-like; Aryl hydrocarbon receptor nuclear translocator-like protein 1; basic-helix-loop-helix-PAS orphan MOP3; Basic-helix-loop-helix-PAS protein MOP3; BHLHE5; bHLH-PAS protein JAP3; Bmal1; Bmal1 protein; BMAL1b; bmal1b'; BMAL1c; BOS_14527; Brain and muscle ARNT-like 1; brain and muscle ARNT-like protein 1; CG8727 PA; Class E basic helix-loop-helix protein 5; cycle; JAP3; Member of PAS protein 3; member of PAS superfamily 3; MGC47515; MOP3; PAS domain-containing protein 3; PASD3; testis tissue sperm-binding protein Li 50e; Tic
Gene Symbols Arntl
Host Species Goat
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11865, 406
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.