Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BMAL1, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABE2627MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE2627MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE2627MI Supplier MilliporeSigma™ Supplier No. ABE2627

Rabbit Polyclonal Antibody

Aryl hydrocarbon receptor nuclear translocator-like protein 1 (UniProt: O00327 also known as Basic-helix-loop-helix-PAS protein MOP3, Brain and muscle ARNT-like 1, BMAL1, Class E basic helix-loop-helix protein 5, bHLHe5; Member of PAS protein 3, PAS domain-containing protein 3, bHLH-PAS protein JAP3) is encoded by the ARNTL (also known as BHLHE5; BMAL1; MOP3; PASD3) gene (gene ID 406) in human. BMLA1 is highly expressed in the adult brain, skeletal muscle, and heart. It is a transcriptional activator that forms a core component of the circadian clock. These components include ARNTL/BMAL1, CLOCK, NPAS2, ARNTL2/BMAL2, PER1, PER2, PER3, CRY1, and CRY2. CLOCK or NPAS2 and ARNTL/BMAL1 or ARNTL2/BMAL2 form the positive limb of the feedback loop. They act in the form of a heterodimer and activate the transcription of core clock genes and clock-controlled genes that are involved in key metabolic processes. PER1/2/3 and CRY1/2, act as transcriptional repressors and form the negative limb of the feedback loop. They interact with the CLOCK|NPAS2-ARNTL/BMAL1|ARNTL2/BMAL2 heterodimer inhibiting its activity. BMLA1 is reported to be phosphorylated upon dimerization with CLOCK and this phosphorylation enhances its transcriptional activity and reduces the stability of the CLOCK-ARNTL/BMAL1 heterodimer by promoting its degradation. BMAL1 is shown to positively regulate myogenesis and negatively regulate adipogenesis via the transcriptional control of the genes of the canonical Wnt signaling pathway. BMAL1 is the only component of the mammalian circadian clock whose deletion in a murine model is shown to generate arrhythmicity. BMAL1 null-mice are reported to have reproductive problems, reduced locomotor activity, and they age quickly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMAL1
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene ARNTL, BHLHE5, BMAL1, MOP3, PASD3
Gene Accession No. O00327
Host Species Rabbit
Immunogen KLH-conjugated peptide corresponding to the C-terminal 20 amino acids of human BMAL1 protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001284648
Test Specificity Detects BMAL1 (Aryl hydrocarbon receptor nuclear translocator-like protein 1) in HeLa cell nuclear extract.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.