Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMAL1 Recombinant Rabbit Monoclonal Antibody (HL2455), Invitrogen™

Catalog No. FEMA550536
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550536 Supplier Thermo Scientific Supplier No. MA550536

Rabbit Recombinant Monoclonal Antibody

The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMAL1
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL2455
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Arntl
Gene Accession No. O00327
Gene Alias Arnt3, ARNTL, ARNT-like protein 1, brain and muscle, aryl hydrocarbon receptor nuclear translocator like, aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, basic-helix-loop-helix-PAS protein MOP3, bHLHe5, bHLH-PAS protein JAP3, BMAL1, Bmal1 protein, BMAL1b, bmal1b', BMAL1c, Brain and muscle ARNT-like 1, brain and muscle Arnt-like protein 1, CG8727 PA, class E basic helix-loop-helix protein 5, cycle, JAP3, LOW QUALITY PROTEIN: aryl hydrocarbon receptor nuclear translocator-like protein 1, member of PAS protein 3, member of PAS superfamily 3, MGC47515, MOP3, PAS domain-containing protein 3, PASD3, testis tissue sperm-binding protein Li 50e, tic
Gene Symbols Arntl
Host Species Rabbit
Immunogen Recombinant fragment of human BMAL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 406
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.