Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bmf Antibody (17A9), Allophycocyanin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280091APC
This item is not returnable.
View return policy
Description
Bmf Monoclonal antibody specifically detects Bmf in Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Bmf | |
Monoclonal | |
APC | |
Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
Recombinant full length mouse Bmf and synthetic peptides corresponding to aa 13-27 and aa 112-126 of mouse Bmf. | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
90427 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry | |
17A9 | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction