Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bmf Antibody (9G10), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280090AF405
This item is not returnable.
View return policy
Description
Bmf Monoclonal antibody specifically detects Bmf in Human, Mouse samples. It is validated for Western Blot, Flow CytometrySpecifications
Bmf | |
Monoclonal | |
Alexa Fluor 405 | |
Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
Recombinant mouse Bmf. | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
90427 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, Flow Cytometry | |
9G10 | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction