Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bmf Antibody (9G10), Allophycocyanin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280090APC
This item is not returnable.
View return policy
Description
Bmf Monoclonal antibody specifically detects Bmf in Human, Mouse samples. It is validated for Flow CytometrySpecifications
| Bmf | |
| Monoclonal | |
| APC | |
| Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
| Recombinant mouse Bmf. | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 90427 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry | |
| 9G10 | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction